missing translation for 'onlineSavingsMsg'
Learn More

Sulfuric Acid, Fuming, puriss. p.a., Solstice

Product Code. 15643110 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15643110 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15643110 Supplier Honeywell Research Chemicals Supplier No. 307361L
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Puriss. p.a., 20% SO3

  • We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories: Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications. Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
TRUSTED_SUSTAINABILITY

Specifications

CAS 8014-95-7
Density 1.925g/mL at 25°C, 1.90g/mL at 20°C
Residue after Ignition ≤0.01%
Vapor Pressure 2mmHg (25°C)
Quantity 1 L
Molecular Formula H2O7S2
Linear Formula H2SO4 · (SO3)x
MDL Number MFCD00137792
UN Number 1831-
Beilstein 2037554
Synonym oleum, fuming sulfuric acid, sulfuric acid, fuming, oleum iodisum, sulfuric acid fuming, oleum fuming sulfuric acid, sulfuric acid, mixt with sulfur trioxide, sulfur trioxide, mixt. with sulfuric acid, sulfuric acid mixture with sulfur trioxide
InChI Key HIFJUMGIHIZEPX-UHFFFAOYSA-N
SMILES O=S(=O)=O.OS(O)(=O)=O
IUPAC Name sulfuric acid;sulfur trioxide
Molecular Weight (g/mol) 178.13
PubChem CID 24681
Formula Weight 98.08g/mol
Grade puriss. p.a.
Chemical Name or Material Sulfuric Acid, Fuming, 20% SO3
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Sulfuric acid, fuming
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 A
  • Specific target organ toxicity Category 3
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H335-May cause respiratory irritation.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-[In case of inadequate ventilation] wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Oleum
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.