accessibility menu, dialog, popup

UserName

CAS RN 81-84-5

OOO

CAS RN 81-84-5

IUPAC Name: 3-oxatricyclo[7.3.1.0⁵,¹³]trideca-1(13),5,7,9,11-pentaene-2,4-dione
Synonyms: 1,8-naphthalic anhydride
Molecular Weight (g/mol): 198.18
Molecular Formula: C12H6O3
InChi Key: GRSMWKLPSNHDHA-UHFFFAOYSA-N
SMILES: O=C1OC(=O)C2=C3C(C=CC=C13)=CC=C2
Molecular Weight (g/mol) 
  • (3)
  • (6)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (6)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (3)