CAS RN 814-95-9
IUPAC Name:
strontium(2+) oxalate
Synonyms:
strontium oxalate
Molecular Weight (g/mol):
175.64
Molecular Formula:
C2O4Sr
InChi Key:
KQAGKTURZUKUCH-UHFFFAOYSA-L
SMILES:
[Sr++].[O-]C(=O)C([O-])=O
1
–
1
of
1
results
1
Strontium oxalate, 95%, Thermo Scientific Chemicals
CAS: 814-95-9 Molecular Formula: C2O4Sr Molecular Weight (g/mol): 175.638 MDL Number: MFCD00050803 InChI Key: KQAGKTURZUKUCH-UHFFFAOYSA-L Synonym: strontium oxalate,unii-7iaw7e69ni,7iaw7e69ni,strontium oxalate, src2o4,ethanedioic acid, strontium salt 1:1,oxalic acid, monostrontium salt,strontium 2+ ion oxalate,oxalic acid, strontium salt 1:1 8ci PubChem CID: 69947 IUPAC Name: strontium;oxalate SMILES: C(=O)(C(=O)[O-])[O-].[Sr+2]