missing translation for 'onlineSavingsMsg'
Learn More

Strontium oxalate, 95%, Thermo Scientific Chemicals

Product Code. 11334477
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11334477 500 g 500g
11324477 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11334477 Supplier Thermo Scientific Alfa Aesar Supplier No. 039573.36
 Restricted Item
Estimated Shipment: 09-09-2026
Add to basket
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 814-95-9
Molecular Formula C2O4Sr
Molecular Weight (g/mol) 175.638
MDL Number MFCD00050803
InChI Key KQAGKTURZUKUCH-UHFFFAOYSA-L
Synonym strontium oxalate, unii-7iaw7e69ni, 7iaw7e69ni, strontium oxalate, src2o4, ethanedioic acid, strontium salt 1:1, oxalic acid, monostrontium salt, strontium 2+ ion oxalate, oxalic acid, strontium salt 1:1 8ci
PubChem CID 69947
IUPAC Name strontium;oxalate
SMILES C(=O)(C(=O)[O-])[O-].[Sr+2]

Specifications

Quantity 500 g
UN Number UN3288
Solubility Information Soluble in hydrochloric acid and nitric acid
Formula Weight 175.64
Percent Purity 95%
Physical Form Powder or Lumps
Chemical Name or Material Strontium oxalate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.