accessibility menu, dialog, popup

UserName

CAS RN 818-38-2

H3COOOOCH3

CAS RN 818-38-2

IUPAC Name: 1,5-diethyl pentanedioate
Synonyms: diethyl glutarate
Molecular Weight (g/mol): 188.22
Molecular Formula: C9H16O4
InChi Key: OUWSNHWQZPEFEX-UHFFFAOYSA-N
SMILES: CCOC(=O)CCCC(=O)OCC
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)