accessibility menu, dialog, popup

UserName

CH3CH3OH3C(E)

CAS RN 81925-81-7

IUPAC Name: (2E)-5-methylhept-2-en-4-one
Synonyms: filbertone
Molecular Weight (g/mol): 126.2
Molecular Formula: C8H14O
InChi Key: ARJWAURHQDJJAC-GQCTYLIANA-N
SMILES: CCC(C)C(=O)\C=C\C
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results