missing translation for 'onlineSavingsMsg'
Learn More

5-Methyl-2-hepten-4-one, predominantly trans, 99%

Product Code. 11456046
Change view
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11456046 10 g 10g
11446046 2 g 2g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11456046 Supplier Thermo Scientific Alfa Aesar Supplier No. B22335.09
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 81925-81-7 | C8H14O | 126.199 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 81925-81-7
Molecular Formula C8H14O
Molecular Weight (g/mol) 126.199
MDL Number MFCD00792509
InChI Key ARJWAURHQDJJAC-GQCTYLIASA-N
Synonym 5-methyl-2-hepten-4-one, filbertone, 5-methylhept-2-en-4-one, +/--filbertone, hazelnut ketone, e-5-methyl-2-hepten-4-one, 5-methyl-e-2-hepten-4-one, fema no. 3761, e-5-methylhept-2-en-4-one, 2e-5-methyl-2-hepten-4-one
PubChem CID 5362588
IUPAC Name (E)-5-methylhept-2-en-4-one
SMILES CCC(C)C(=O)C=CC

Specifications

Boiling Point 169°C to 171°C
Flash Point 55°C (131°F)
Refractive Index 1.444
Quantity 10 g
UN Number UN1224
Formula Weight 126.2
Percent Purity 99%
Chemical Name or Material 5-Methyl-2-hepten-4-one, predominantly trans

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.