accessibility menu, dialog, popup

UserName

CAS RN 82380-18-5

OHNF

CAS RN 82380-18-5

IUPAC Name: 2-fluoro-4-hydroxybenzonitrile
Synonyms: 2-fluoro-4-hydroxybenzonitrile
Molecular Weight (g/mol): 137.11
Molecular Formula: C7H4FNO
InChi Key: REIVHYDACHXPNH-UHFFFAOYSA-N
SMILES: OC1=CC=C(C#N)C(F)=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results