accessibility menu, dialog, popup

UserName

CAS RN 824-45-3

CH3CH3Cl

CAS RN 824-45-3

IUPAC Name: 2-(chloromethyl)-1,4-dimethylbenzene
Synonyms: 2-(chloromethyl)-1,4-dimethylbenzene
Molecular Weight (g/mol): 154.64
Molecular Formula: C9H11Cl
InChi Key: PECXPZGFZFGDRD-UHFFFAOYSA-N
SMILES: CC1=CC=C(C)C(CCl)=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results