accessibility menu, dialog, popup

UserName

OHClOHCl

CAS RN 824-69-1

IUPAC Name: 2,5-dichlorobenzene-1,4-diol
Synonyms: 2,5-dichlorohydroquinone 2,5-dichloro-1,4-hydroquinone 2,5-dichloro-p-hydroquinone
Molecular Weight (g/mol): 179
Molecular Formula: C6H4Cl2O2
InChi Key: AYNPIRVEWMUJDE-UHFFFAOYSA-N
SMILES: OC1=CC(Cl)=C(O)C=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results