accessibility menu, dialog, popup

UserName

ONO

CAS RN 82413-63-6

IUPAC Name: 4-[(morpholin-4-yl)methyl]benzaldehyde
Synonyms: 4-(morpholin-4-ylmethyl)benzaldehyde
Molecular Weight (g/mol): 205.26
Molecular Formula: C12H15NO2
InChi Key: KMAHWHPUXGNVBN-UHFFFAOYSA-N
SMILES: O=CC1=CC=C(CN2CCOCC2)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results