accessibility menu, dialog, popup

UserName

CAS RN 82586-52-5

ClH3COO(S)NH(S)H3CONOCH3OH3C(S)HOOH

CAS RN 82586-52-5

IUPAC Name: hydrogen (3S)-2-[(2S)-2-{[(2S)-1-ethoxy-1-oxo-4-phenylbutan-2-yl]amino}propanoyl]-6,7-dimethoxy-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid chloride
Synonyms: hydrogen moexipril chloride
Molecular Weight (g/mol): 535.03
Molecular Formula: C27H35ClN2O7
InChi Key: JXRAXHBVZQZSIC-JKVLGAQCSA-N
SMILES: [H+].[Cl-].CCOC(=O)[C@H](CCC1=CC=CC=C1)N[C@@H](C)C(=O)N1CC2=CC(OC)=C(OC)C=C2C[C@H]1C(O)=O
Grade 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results