missing translation for 'onlineSavingsMsg'
Learn More

Moexipril hydrochloride, MedChemExpress

Product Code. 16498266
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
This item is not returnable. View return policy

Product Code. 16498266

Brand: Medchem Express HYB0378A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Moexipril hydrochloride (RS-10085) is an orally active inhibitor of angiotensin-converting enzyme (ACE), and becomes effective by being hydrolyzed to moexiprila (hydrochloride). Moexipril hydrochloride exhibits antihypertensive and neuroprotective effects-.

Moexipril hydrochloride (RS-10085) is an orally active inhibitor of angiotensin-converting enzyme (ACE), and becomes effective by being hydrolyzed to moexiprila (hydrochloride). Moexipril hydrochloride exhibits antihypertensive and neuroprotective effects-.

Specifications

Chemical Name or Material Moexipril hydrochloride
CAS 82586-52-5
Color White
Purity Grade Notes Research
Molecular Formula C27H35ClN2O7
Quantity 100 mg
Synonym RS-10085
Solubility Information DMSO : 250 mg/mL (467.26 mM; Need ultrasonic) ∣H2O : 100 mg/mL (186.91 mM; Need ultrasonic)
SMILES O=C([C@H]1N(C([C@@H](N[C@H](C(OCC)=O)CCC2=CC=CC=C2)C)=O)CC3=C(C=C(OC)C(OC)=C3)C1)O.Cl
Molecular Weight (g/mol) 535.03
Formula Weight 535.03
Percent Purity 98.09%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.