accessibility menu, dialog, popup

UserName

H2C

CAS RN 826-74-4

IUPAC Name: 1-ethenylnaphthalene
Synonyms: 1-vinylnaphthalene
Molecular Weight (g/mol): 154.21
Molecular Formula: C12H10
InChi Key: IGGDKDTUCAWDAN-UHFFFAOYSA-N
SMILES: C=CC1=C2C=CC=CC2=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)