missing translation for 'onlineSavingsMsg'
Learn More

1-Vinylnaphthalene [Stabilized with 4000ppm tert-Butylcatechol], TRC

Product Code. 30450366
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30450366 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30450366 Supplier Toronto Research Chemicals Supplier No. TRCV4250005G
 Restricted Item
Estimated Shipment: 29-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifica

Chemical Name or Material 1-Vinylnaphthalene
CAS 826-74-4
Molecular Formula C12 H10
Quantity 5 g
Synonym 1-Vinylnaphthalene
InChI Formula InChI=1S/C12H10/c1-2-10-7-5-8-11-6-3-4-9-12(10)11/h2-9H,1H2
SMILES C=Cc1cccc2ccccc12
IUPAC Name 1-ethenylnaphthalene
Molecular Weight (g/mol) 154.21
Formula Weight 154.0783
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.