accessibility menu, dialog, popup

UserName

CH2

CAS RN 827-54-3

IUPAC Name: 2-ethenylnaphthalene
Synonyms: 2-vinylnaphthalene
Molecular Weight (g/mol): 154.21
Molecular Formula: C12H10
InChi Key: KXYAVSFOJVUIHT-UHFFFAOYSA-N
SMILES: C=CC1=CC=C2C=CC=CC2=C1
Boiling Point 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)