missing translation for 'onlineSavingsMsg'
Learn More

2-Vinylnaphthalene, 97%

Product Code. 11454074
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11454074 25 g 25g
11444074 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11454074 Supplier Thermo Scientific Alfa Aesar Supplier No. A12470.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 827-54-3 | C12H10 | 154.21 g/mol

2-Vinylnaphthalene is employed in the synthesis of various copolymers such as Poly (sodium styrenesulfonate-co-2-vinyl-napthalene) copolymer and Poly (2-vinylnapthalene-alt-maleic acid)-graft-polystyrene. The latter finds applications as a photoactive polymer micelle and stabilizer for polystyrene latexes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Vinylnaphthalene is employed in the synthesis of various copolymers such as Poly (sodium styrenesulfonate-co-2-vinyl-napthalene) copolymer and Poly (2-vinylnapthalene-alt-maleic acid)-graft-polystyrene. The latter finds applications as a photoactive polymer micelle and stabilizer for polystyrene latexes.

Solubility
Soluble in ethanol, acetone, and benzene. Insoluble in water.

Notes
Store under cool dry place. Ensure good ventilation. Incompatible with oxidizing agents. Protect away from heat.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 827-54-3
Molecular Formula C12H10
Molecular Weight (g/mol) 154.21
MDL Number MFCD00004125
InChI Key KXYAVSFOJVUIHT-UHFFFAOYSA-N
Synonym 2-vinylnaphthalene, naphthalene, 2-ethenyl, 2-vinyl-naphthalene, beta-vinylnaphthalene, poly 2-vinylnaphthalene, unii-hzd8li91n1, hzd8li91n1, polyvinylnaphthalene, 2-vinyl naphthalene, 2-ethenyinaphthalene
PubChem CID 13230
ChEBI CHEBI:51325
IUPAC Name 2-ethenylnaphthalene
SMILES C=CC1=CC=C2C=CC=CC2=C1

Specifications

Melting Point 62°C to 67°C
Boiling Point 128°C to 131°C (13 mmHg)
Flash Point >110°C (230°F)
Quantity 25 g
Beilstein 2038190
Solubility Information Soluble in ethanol,acetone,and benzene. Insoluble in water.
Formula Weight 154.21
Percent Purity 97%
Chemical Name or Material 2-Vinylnaphthalene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.