accessibility menu, dialog, popup

UserName

SFFFFFOHO

CAS RN 833-96-5

IUPAC Name: 3-(pentafluoro-λ⁶-sulfanyl)benzoic acid
Synonyms: 3-(pentafluoro-λ⁶-sulfanyl)benzoic acid
Molecular Weight (g/mol): 248.17
Molecular Formula: C7H5F5O2S
InChi Key: AKFDIYXSRGLRAB-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC(=CC=C1)S(F)(F)(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results