accessibility menu, dialog, popup

UserName

CAS RN 833472-82-5

BOOCH3HOOH

CAS RN 833472-82-5

IUPAC Name: [3-(3-methoxy-3-oxopropyl)phenyl]boronic acid
Synonyms: 3-(3-methoxy-3-oxopropyl)phenylboronic acid
Molecular Weight (g/mol): 208.02
Molecular Formula: C10H13BO4
InChi Key: IPXZIWCEVNDHFD-UHFFFAOYSA-N
SMILES: COC(=O)CCC1=CC=CC(=C1)B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results