missing translation for 'onlineSavingsMsg'
Learn More

3-(2-Methoxycarbonylethyl)benzeneboronic acid, 97%

Product Code. 15469277
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15469277 1 g 1g
15459895 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15469277 Supplier Thermo Scientific Alfa Aesar Supplier No. H52542.03
 Restricted Item
Estimated Shipment: 03-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 833472-82-5 | C10H13BO4 | 208.02 g/mol

3-(2-Methoxycarbonylethyl)benzeneboronic acid is involved in the preparation of antimalarial and anticancer ethylenediamine inhibitors to protein farnesyltransferase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-(2-Methoxycarbonylethyl)benzeneboronic acid is involved in the preparation of antimalarial and anticancer ethylenediamine inhibitors to protein farnesyltransferase.

Notes
Incompatible with strong oxidizing agents and strong acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 833472-82-5
Molecular Formula C10H13BO4
Molecular Weight (g/mol) 208.02
MDL Number MFCD04115648
InChI Key IPXZIWCEVNDHFD-UHFFFAOYSA-N
Synonym 3-2-methoxycarbonylethyl phenylboronic acid, methyl 3-3-boronophenyl propionate, 3-3-methoxy-3-oxopropyl phenyl boronic acid, 3-3-methoxy-3-oxopropyl phenylboronic acid, 3-2-methoxycarbonylethyl phenyl boronic acid, acmc-209prx, benzenepropanoic acid,3-borono-, 1-methyl ester
PubChem CID 4993880
IUPAC Name [3-(3-methoxy-3-oxopropyl)phenyl]boronic acid
SMILES COC(=O)CCC1=CC=CC(=C1)B(O)O

Specifications

Melting Point 96°C to 100°C
Quantity 1 g
Formula Weight 208.02
Percent Purity 97%
Chemical Name or Material 3-(2-Methoxycarbonylethyl)benzeneboronic Acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.