CAS RN 83846-83-7
CAS RN 83846-83-7
IUPAC Name:
2,3-dihydroxybutanedioic acid 3-{2-[4-(4-fluorobenzoyl)piperidin-1-yl]ethyl}-1,2,3,4-tetrahydroquinazoline-2,4-dione hydrate
Synonyms:
(.+-.)-tartaric acid ketanserin hydrate
Molecular Weight (g/mol):
563.54
Molecular Formula:
C26H30FN3O10
InChi Key:
KJJRKCWDAHTVRL-UHFFFAOYNA-N
SMILES:
O.OC(C(O)C(O)=O)C(O)=O.FC1=CC=C(C=C1)C(=O)C1CCN(CCN2C(=O)NC3=CC=CC=C3C2=O)CC1
1
–
3
of
3
results
2
Tocris Bioscience™ Ketanserin tartrate
CAS: 83846-83-7 Molecular Formula: C26H30FN3O10
3
Ketanserin tartrate, MedChemExpress
MedChemExpress Ketanserin (R41468) tartrate is a selective 5-HT2 receptor antagonist. Ketanserin tartrate also blocks hERG current (IhERG) in a concentration-dependent manner (IC50=0.11 μM).