accessibility menu, dialog, popup

UserName

OO

CAS RN 84-11-7

IUPAC Name: 9,10-dihydrophenanthrene-9,10-dione
Synonyms: 9,10-phenanthrenequinone phenanthraquinone phenanthrenequinone 9,10-phenanthroquinone 9,10-phenanthraquinone 9-10 phenanthrene quinone
Molecular Weight (g/mol): 208.22
Molecular Formula: C14H8O2
InChi Key: YYVYAPXYZVYDHN-UHFFFAOYSA-N
SMILES: O=C1C(=O)C2=CC=CC=C2C2=CC=CC=C12
Molecular Weight (g/mol) 
  • (6)
  • (3)
  • (2)
Percent Purity 
  • (5)
  • (6)
Quantity 
  • (1)
  • (1)
  • (3)
  • (3)
  • (2)
  • (1)
Boiling Point 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (3)
  • (6)
  • (6)
  • (3)
  • (2)
  • (5)
  • (6)
  • (1)
  • (1)
  • (3)
  • (3)
  • (2)
  • (1)
  • (3)
  • (2)
  • (2)