accessibility menu, dialog, popup

UserName

CH3OO

CAS RN 84-51-5

IUPAC Name: 2-ethyl-9,10-dihydroanthracene-9,10-dione
Synonyms: 2-ethylanthraquinone
Molecular Weight (g/mol): 236.27
Molecular Formula: C16H12O2
InChi Key: SJEBAWHUJDUKQK-UHFFFAOYSA-N
SMILES: CCC1=CC=C2C(=O)C3=CC=CC=C3C(=O)C2=C1
Quantity 
  • (1)
  • (1)
  • (3)
  • (1)
Percent Purity 
  • (1)
  • (3)
Boiling Point 
  • (3)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (3)
  • (3)
  • (1)