accessibility menu, dialog, popup

UserName

CAS RN 84-89-9

NH2SOHOO

CAS RN 84-89-9

IUPAC Name: 5-aminonaphthalene-1-sulfonic acid
Synonyms: 5-aminonaphthalene-1-sulfonic acid
Molecular Weight (g/mol): 223.25
Molecular Formula: C10H9NO3S
InChi Key: DQNAQOYOSRJXFZ-UHFFFAOYSA-N
SMILES: NC1=C2C=CC=C(C2=CC=C1)S(O)(=O)=O
Quantity 
  • (2)
  • (2)
  • (1)
  • (1)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (3)