accessibility menu, dialog, popup

UserName

CH3NNOHOBr

CAS RN 84547-86-4

IUPAC Name: 4-bromo-1-methyl-1H-pyrazole-3-carboxylic acid
Synonyms: 4-bromo-1-methylpyrazole-3-carboxylic acid
Molecular Weight (g/mol): 205.01
Molecular Formula: C5H5BrN2O2
InChi Key: LEEPGDCCHVRYHK-UHFFFAOYSA-N
SMILES: CN1C=C(Br)C(=N1)C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)