missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-1-methyl-1H-pyrazole-3-carboxylic acid, 97%, Thermo Scientific™

Product Code. 10443214
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10443214 1 g 1g
10327364 250 mg 250mg
10178224 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10443214 Supplier Thermo Scientific Maybridge Supplier No. CC63901DA
 Restricted Item
Estimated Shipment: 05-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 84547-86-4
Molecular Formula C5H5BrN2O2
Molecular Weight (g/mol) 205.011
InChI Key LEEPGDCCHVRYHK-UHFFFAOYSA-N
Synonym 4-bromo-1-methyl-1h-pyrazole-3-carboxylic acid, 1h-pyrazole-3-carboxylic acid, 4-bromo-1-methyl, 4-bromo-1-methyl-1h-pyrazole-3-carboxylicacid, pubchem16814, 4-bromo-1-methyl-pyrazole-3-carboxylic acid, 1h-pyrazole-3-carboxylicacid, 4-bromo-1-methyl, 4-bromo-1-methyl-1h-pyrazole-3-carboxylic acid #
PubChem CID 536013
IUPAC Name 4-bromo-1-methylpyrazole-3-carboxylic acid
SMILES CN1C=C(C(=N1)C(=O)O)Br

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 205.01
Percent Purity 97%
Chemical Name or Material 4-Bromo-1-methyl-1H-pyrazole-3-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.