accessibility menu, dialog, popup

UserName

CAS RN 85-56-3

HOOOCl

CAS RN 85-56-3

IUPAC Name: 2-(4-chlorobenzoyl)benzoic acid
Synonyms: 2-(4-chlorobenzoyl)benzoic acid
Molecular Weight (g/mol): 260.67
Molecular Formula: C14H9ClO3
InChi Key: YWECCEXWKFHHQJ-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=CC=C1C(=O)C1=CC=C(Cl)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results