accessibility menu, dialog, popup

UserName

CAS RN 850568-00-2

IUPAC Name: (4-fluoro-2-hydroxyphenyl)boronic acid
Synonyms: 4-fluoro-2-hydroxyphenylboronic acid
Molecular Weight (g/mol): 155.92
Molecular Formula: C6H6BFO3
InChi Key: KOFNQIAWWVBBJZ-UHFFFAOYSA-N
SMILES: OB(O)C1=C(O)C=C(F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results