accessibility menu, dialog, popup

UserName

BNHOOH(E/Z)

CAS RN 850568-53-5

IUPAC Name: [3-(2-cyanoeth-1-en-1-yl)phenyl]boronic acid
Synonyms: 3-(2-cyanoeth-1-en-1-yl)phenylboronic acid
Molecular Weight (g/mol): 172.98
Molecular Formula: C9H8BNO2
InChi Key: YDSLGHLJEJPWNC-UHFFFAOYSA-N
SMILES: OB(O)C1=CC(C=CC#N)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results