accessibility menu, dialog, popup

UserName

CAS RN 850568-80-8

BClOCH3HOOH

CAS RN 850568-80-8

IUPAC Name: (4-chloro-2-ethoxyphenyl)boronic acid
Synonyms: 4-chloro-2-ethoxyphenylboronic acid
Molecular Weight (g/mol): 200.43
Molecular Formula: C8H10BClO3
InChi Key: NIWJJDHZRLYASL-UHFFFAOYSA-N
SMILES: CCOC1=C(C=CC(Cl)=C1)B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results