CAS RN 850623-64-2
IUPAC Name:
potassium trifluoro(2-nitrophenyl)boranuide
Synonyms:
potassium trifluoro(2-nitrophenyl)boranuide
Molecular Weight (g/mol):
229.01
Molecular Formula:
C6H4BF3KNO2
InChi Key:
DZHYOFKWMHZKBK-UHFFFAOYSA-N
SMILES:
[K+].[O-][N+](=O)C1=CC=CC=C1[B-](F)(F)F
1
–
1
of
1
results
1
Potassium (2-nitrophenyl)trifluoroborate, 97%, Thermo Scientific™
CAS: 850623-64-2 Molecular Formula: C6H4BF3KNO2 Molecular Weight (g/mol): 229.01 InChI Key: DZHYOFKWMHZKBK-UHFFFAOYSA-N Synonym: potassium 2-nitrophenyl trifluoroborate,potassium trifluoro 2-nitrophenyl borate,potassium trifluoro 2-nitrophenyl boranuide,potassium trifluoro-2-nitrophenyl boranuide,potassium 2-nitrophenyltrifluoroborate,amtb936,potassium ion trifluoro 2-nitrophenyl boranuide,potassium tris fluoranyl-2-nitrophenyl boranuide PubChem CID: 2782854 IUPAC Name: potassium;trifluoro-(2-nitrophenyl)boranuide SMILES: [B-](C1=CC=CC=C1[N+](=O)[O-])(F)(F)F.[K+]