missing translation for 'onlineSavingsMsg'
Learn More

Potassium (2-nitrophenyl)trifluoroborate, 97%, Thermo Scientific™

Product Code. 11946081 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11946081 5 g 5g
11926081 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11946081 Supplier Thermo Scientific Acros Supplier No. 444850050
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 850623-64-2
Molecular Formula C6H4BF3KNO2
Molecular Weight (g/mol) 229.01
InChI Key DZHYOFKWMHZKBK-UHFFFAOYSA-N
Synonym potassium 2-nitrophenyl trifluoroborate, potassium trifluoro 2-nitrophenyl borate, potassium trifluoro 2-nitrophenyl boranuide, potassium trifluoro-2-nitrophenyl boranuide, potassium 2-nitrophenyltrifluoroborate, amtb936, potassium ion trifluoro 2-nitrophenyl boranuide, potassium tris fluoranyl-2-nitrophenyl boranuide
PubChem CID 2782854
IUPAC Name potassium;trifluoro-(2-nitrophenyl)boranuide
SMILES [B-](C1=CC=CC=C1[N+](=O)[O-])(F)(F)F.[K+]

Specifications

Melting Point 226°C to 228°C
Assay Percent Range 97%
Quantity 5 g
Formula Weight 229.01
Percent Purity 97%
Chemical Name or Material Potassium (2-nitrophenyl)trifluoroborate
compliance-icons
Identifiant produit
  • Potassium (2-nitrophenyl)trifluoroborate
Mention d'avertissement
  • Warning
Catégorie de danger
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Mention de danger
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Conseils de prudence
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.