accessibility menu, dialog, popup

UserName

CAS RN 850623-75-5

BSCH3FFFK

CAS RN 850623-75-5

IUPAC Name: potassium [4-(ethylsulfanyl)phenyl]trifluoroboranuide
Synonyms: potassium [4-(ethylsulfanyl)phenyl]trifluoroboranuide
Molecular Weight (g/mol): 244.12
Molecular Formula: C8H9BF3KS
InChi Key: BVLNLNRSQDZWIL-UHFFFAOYSA-N
SMILES: [K+].CCSC1=CC=C(C=C1)[B-](F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results