accessibility menu, dialog, popup

UserName

CAS RN 85068-28-6

OHOFF

CAS RN 85068-28-6

IUPAC Name: 2-(2,6-difluorophenyl)acetic acid
Synonyms: (2,6-difluorophenyl)acetic acid
Molecular Weight (g/mol): 172.13
Molecular Formula: C8H6F2O2
InChi Key: FUGDCKXBUZFEON-UHFFFAOYSA-N
SMILES: OC(=O)CC1=C(F)C=CC=C1F
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results