accessibility menu, dialog, popup

UserName

CAS RN 852228-15-0

IUPAC Name: 4-benzylpiperazine-1-carboximidamide hydroiodide
Synonyms: 4-benzylpiperazine-1-carboximidamide hydroiodide
Molecular Weight (g/mol): 346.22
Molecular Formula: C12H19IN4
InChi Key: CXQITJKGJPFOIV-UHFFFAOYSA-N
SMILES: I.NC(=N)N1CCN(CC2=CC=CC=C2)CC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results