accessibility menu, dialog, popup

UserName

NHNCl(E/Z)

CAS RN 852691-01-1

IUPAC Name: 5-[2-(4-chlorophenyl)ethenyl]-1H-pyrazole
Synonyms: 3-[2-(4-chlorophenyl)ethenyl]-2H-pyrazole
Molecular Weight (g/mol): 204.66
Molecular Formula: C11H9ClN2
InChi Key: SHJXJPOMWYIJSA-UHFFFAOYSA-N
SMILES: ClC1=CC=C(C=CC2=CC=NN2)C=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results