accessibility menu, dialog, popup

UserName

CAS RN 85466-66-6

H3CH3CCH3OONCH3OHO

CAS RN 85466-66-6

IUPAC Name: 2-{[(tert-butoxy)carbonyl](methyl)amino}-3-phenylpropanoic acid
Synonyms: 2-[(tert-butoxycarbonyl)(methyl)amino]-3-phenylpropanoic acid
Molecular Weight (g/mol): 279.34
Molecular Formula: C15H21NO4
InChi Key: AJGJINVEYVTDNH-UHFFFAOYNA-N
SMILES: CN(C(CC1=CC=CC=C1)C(O)=O)C(=O)OC(C)(C)C
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)