accessibility menu, dialog, popup

UserName

HNNH2SOOOHH2O

CAS RN 854689-07-9

IUPAC Name: 4-hydrazinylbenzene-1-sulfonic acid hydrate
Synonyms: 4-hydrazinylbenzenesulfonic acid hydrate
Molecular Weight (g/mol): 206.22
Molecular Formula: C6H10N2O4S
InChi Key: OSOMMFVBTYFMBI-UHFFFAOYSA-N
SMILES: O.NNC1=CC=C(C=C1)S(O)(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results