accessibility menu, dialog, popup

UserName

ONOHO

CAS RN 857284-07-2

IUPAC Name: {3-[2-(morpholin-4-yl)ethoxy]phenyl}methanol
Synonyms: {3-[2-(morpholin-4-yl)ethoxy]phenyl}methanol
Molecular Weight (g/mol): 237.3
Molecular Formula: C13H19NO3
InChi Key: DZHYOLUKMGNRTO-UHFFFAOYSA-N
SMILES: OCC1=CC(OCCN2CCOCC2)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results