CAS RN 85798-08-9
CAS RN 85798-08-9
IUPAC Name:
hydrogen (4aR,8aR)-5-propyl-1H,4H,4aH,5H,6H,7H,8H,8aH,9H-pyrido[2,3-f]indazole chloride
Synonyms:
hydrogen quinpirole chloride
Molecular Weight (g/mol):
255.79
Molecular Formula:
C13H22ClN3
InChi Key:
HJHVRVJTYPKTHX-HTMVYDOJSA-N
SMILES:
[H+].[Cl-].CCCN1CCC[C@@H]2CC3=C(C[C@@H]12)C=NN3
1
–
2
of
2
results
1
(-)-Quinpirole Hydrochloride, TRC
CAS: 85798-08-9 Molecular Formula: C13H21N3 . (HCl) Molecular Weight (g/mol): 255.79 Synonym: (4aR,8aR)-4,4a,5,6,7,8,8a,9-Octahydro-5-propyl-1H-pyrazolo[3,4-g]quinoline Monohydrochloride,(-)-LY 171555,LY 171555,Quinpirole Hydrochloride,(4aR-trans)-4,4a,5,6,7,8,8a,9-Octahydro-5-propyl-1H-pyrazolo[3,4-g]quinoline Monohydrochloride IUPAC Name: (4aR,8aR)-5-propyl-1,4,4a,6,7,8,8a,9-octahydropyrazolo[3,4-g]quinoline;hydrochloride SMILES: Cl.CCCN1CCC[C@@H]2Cc3[nH]ncc3C[C@@H]12
2
Promotions Available
Tocris Bioscience™ (-)-Quinpirole hydrochloride
Tocris Bioscience™ (-)-Quinpirole hydrochloride
CAS: 85798-08-9 Molecular Formula: C13H22ClN3