missing translation for 'onlineSavingsMsg'
Learn More

(-)-Quinpirole Hydrochloride, TRC

Product Code. 30418033
Change view
Click to view available options
Quantity:
1 mg
2.5 mg
5 mg
Unit Size:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30418033 2.5 mg 2.5mg
30417223 1 mg -
30409948 5 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30418033 Supplier Toronto Research Chemicals Supplier No. TRCQ7967132.5MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 85798-08-9
Molecular Formula C13H21N3 . (HCl)
Molecular Weight (g/mol) 255.79
Synonym (4aR,8aR)-4,4a,5,6,7,8,8a,9-Octahydro-5-propyl-1H-pyrazolo[3,4-g]quinoline Monohydrochloride, (-)-LY 171555, LY 171555, Quinpirole Hydrochloride, (4aR-trans)-4,4a,5,6,7,8,8a,9-Octahydro-5-propyl-1H-pyrazolo[3,4-g]quinoline Monohydrochloride
IUPAC Name (4aR,8aR)-5-propyl-1,4,4a,6,7,8,8a,9-octahydropyrazolo[3,4-g]quinoline;hydrochloride
SMILES Cl.CCCN1CCC[C@@H]2Cc3[nH]ncc3C[C@@H]12

Specifications

Quantity 2.5 mg
InChI Formula InChI=1S/C13H21N3.ClH/c1-2-5-16-6-3-4-10-7-12-11(8-13(10)16)9-14-15-12;/h9-10,13H,2-8H2,1H3,(H,14,15);1H/t10-,13-;/m1./s1
Formula Weight 255.1502
Chemical Name or Material (-)-Quinpirole Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.