CAS RN 858839-90-4
CAS RN 858839-90-4
IUPAC Name:
4-iodo-1,2-dihydropyridin-2-one
Synonyms:
4-iodo-1H-pyridin-2-one
Molecular Weight (g/mol):
221
Molecular Formula:
C5H4INO
InChi Key:
POHJRJNCZQENQG-UHFFFAOYSA-N
SMILES:
IC1=CC(=O)NC=C1
1
–
1
of
1
results
1
4-Iodoopyridin-2(1H)-one, 97+%, Thermo Scientific™
CAS: 858839-90-4 Molecular Formula: C5H4INO Molecular Weight (g/mol): 221.00 MDL Number: MFCD09702419 InChI Key: POHJRJNCZQENQG-UHFFFAOYSA-N Synonym: 4-iodopyridin-2 1h-one,4-iodo-2-pyridone,4-iodopyridin-2-ol,4-iodo-1,2-dihydropyridin-2-one,2 1h-pyridinone,4-iodo,2-hydroxy-4-iodopyridine,4-iodohydropyridin-2-one,4-iodo-pyridin-2-ol PubChem CID: 20582535 SMILES: IC1=CC(=O)NC=C1