missing translation for 'onlineSavingsMsg'
Learn More

4-Iodoopyridin-2(1H)-one, 97+%, Thermo Scientific™

Product Code. 10694385
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 10694385

Brand: Thermo Scientific Maybridge MO07919CB

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 858839-90-4
Molecular Formula C5H4INO
Molecular Weight (g/mol) 221.00
MDL Number MFCD09702419
InChI Key POHJRJNCZQENQG-UHFFFAOYSA-N
Synonym 4-iodopyridin-2 1h-one, 4-iodo-2-pyridone, 4-iodopyridin-2-ol, 4-iodo-1,2-dihydropyridin-2-one, 2 1h-pyridinone,4-iodo, 2-hydroxy-4-iodopyridine, 4-iodohydropyridin-2-one, 4-iodo-pyridin-2-ol
PubChem CID 20582535
SMILES IC1=CC(=O)NC=C1

Specifications

Assay Percent Range 97+%
Packaging Amber Glass Bottle
Quantity 250 mg
IUPAC Name 4-iodo-1,2-dihydropyridin-2-one
Formula Weight 220.99
Percent Purity 97%
Chemical Name or Material 4-iodo-2-pyridone
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.