accessibility menu, dialog, popup

UserName

ClSClOOCl

CAS RN 85952-30-3

IUPAC Name: (3,4-dichlorophenyl)methanesulfonyl chloride
Synonyms: (3,4-dichlorophenyl)methanesulfonyl chloride
Molecular Weight (g/mol): 259.53
Molecular Formula: C7H5Cl3O2S
InChi Key: DRYGYFAFCNDLEJ-UHFFFAOYSA-N
SMILES: ClC1=CC=C(CS(Cl)(=O)=O)C=C1Cl
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (1)