missing translation for 'onlineSavingsMsg'
Learn More

(3,4-Dichlorophenyl)methanesulfonyl chloride, 90%, Thermo Scientific™

Product Code. 10514373
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10514373 1 g 1g
10064582 250 mg 250mg
10198824 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10514373 Supplier Thermo Scientific Maybridge Supplier No. MO00820DA
 Restricted Item
Estimated Shipment: 05-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 85952-30-3
Molecular Formula C7H5Cl3O2S
Molecular Weight (g/mol) 259.525
MDL Number MFCD01631929
InChI Key DRYGYFAFCNDLEJ-UHFFFAOYSA-N
Synonym 3,4-dichlorophenyl methanesulfonyl chloride, 3,4-dichloro-phenyl-methanesulfonyl chloride, 3,4-dichlorophenyl methanesulphonyl chloride, 3,4-dichlorophenyl methyl sulfonyl chloride, 3,4-dichlorobenzylsulfonyl chloride, 3,4-dichlorophenyl methyl chlorosulfone, 3,4-dichloro-alpha-toluenesulfonyl chloride, 3,4-dichlorophenyl-methanesulfonyl chloride, benzenemethanesulfonyl chloride, 3,4-dichloro, 3,4-dichlorophenyl methyl sulphonyl chloride
PubChem CID 2794622
IUPAC Name (3,4-dichlorophenyl)methanesulfonyl chloride
SMILES C1=CC(=C(C=C1CS(=O)(=O)Cl)Cl)Cl

Specifications

CAS Min % 90.0
Assay Percent Range 90%
Packaging Amber glass bottle
Quantity 1 g
Formula Weight 259.54
Percent Purity 90%
Chemical Name or Material (3, 4-Dichlorophenyl)methanesulfonyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.