accessibility menu, dialog, popup

UserName

NOH

CAS RN 859850-72-9

IUPAC Name: [3-(pyrrolidin-1-yl)phenyl]methanol
Synonyms: [3-(pyrrolidin-1-yl)phenyl]methanol
Molecular Weight (g/mol): 177.25
Molecular Formula: C11H15NO
InChi Key: MYIYSGIMXUQECR-UHFFFAOYSA-N
SMILES: OCC1=CC(=CC=C1)N1CCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results