missing translation for 'onlineSavingsMsg'
Learn More

(3-Pyrrolidin-1-ylphenyl)methanol, 97%, Thermo Scientific™

Product Code. 10107772
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10107772 1 g 1g
10045852 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10107772 Supplier Thermo Scientific Maybridge Supplier No. CC38509DA
 Restricted Item
Estimated Shipment: 12-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 859850-72-9
Molecular Formula C11H15NO
Molecular Weight (g/mol) 177.247
MDL Number MFCD07772826
InChI Key MYIYSGIMXUQECR-UHFFFAOYSA-N
Synonym 3-pyrrolidin-1-ylphenyl methanol, 3-pyrrolidin-1-yl phenyl methanol, 3-1-pyrrolidinyl benzyl alcohol, 3-pyrrolidinylphenyl methan-1-ol, benzenemethanol,3-1-pyrrolidinyl
PubChem CID 7162062
IUPAC Name (3-pyrrolidin-1-ylphenyl)methanol
SMILES C1CCN(C1)C2=CC=CC(=C2)CO

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1 g
Formula Weight 177.25
Percent Purity 97%
Chemical Name or Material (3-Pyrrolidin-1-ylphenyl)methanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.