CAS RN 860644-28-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Losartan Isomer Impurity, Potassium Salt, TRC
CAS: 860644-28-6 Molecular Formula: C22H22ClKN6O Molecular Weight (g/mol): 461 Synonym: 2-Butyl-5-chloro-1-[[2'-(2H-tetrazol-5-yl)[1,1'-biphenyl]-4-yl]methyl]-1H-imidazole-4-methanol Potassium Salt,Losartan Isomer Impurity,Losartan EP Impurity C,Isolosartan IUPAC Name: potassium;[2-butyl-5-chloro-1-[[4-[2-(1,2,3-triaza-4-azanidacyclopenta-2,5-dien-5-yl)phenyl]phenyl]methyl]imidazol-4-yl]methanol SMILES: OCC1=C(Cl)N(CC(C=C2)=CC=C2C3=C(C4=N[N-]N=N4)C=CC=C3)C(CCCC)=N1.[K+]