missing translation for 'onlineSavingsMsg'
Learn More

Losartan Isomer Impurity, Potassium Salt, TRC

Product Code. 30134676
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30134676 5 mg 5mg
30008132 10 mg 10mg
30011297 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134676 Supplier Toronto Research Chemicals Supplier No. TRCL4705255MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 860644-28-6
Molecular Formula C22H22ClKN6O
Molecular Weight (g/mol) 461
Synonym 2-Butyl-5-chloro-1-[[2'-(2H-tetrazol-5-yl)[1,1'-biphenyl]-4-yl]methyl]-1H-imidazole-4-methanol Potassium Salt, Losartan Isomer Impurity, Losartan EP Impurity C, Isolosartan
IUPAC Name potassium;[2-butyl-5-chloro-1-[[4-[2-(1,2,3-triaza-4-azanidacyclopenta-2,5-dien-5-yl)phenyl]phenyl]methyl]imidazol-4-yl]methanol
SMILES OCC1=C(Cl)N(CC(C=C2)=CC=C2C3=C(C4=N[N-]N=N4)C=CC=C3)C(CCCC)=N1.[K+]

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C22H22ClN6O.K/c1-2-3-8-20-24-19(14-30)21(23)29(20)13-15-9-11-16(12-10-15)17-6-4-5-7-18(17)22-25-27-28-26-22;/h4-7,9-12,30H,2-3,8,13-14H2,1H3;/q-1;+1
Formula Weight 460.1181
Percent Purity >95%
Chemical Name or Material Losartan Isomer Impurity, Potassium Salt
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.