accessibility menu, dialog, popup

UserName

HOO(S)NOO

CAS RN 86069-86-5

IUPAC Name: (2S)-1-{[(9H-fluoren-9-yl)methoxy]carbonyl}piperidine-2-carboxylic acid
Synonyms: (2S)-1-[(9H-fluoren-9-ylmethoxy)carbonyl]piperidine-2-carboxylic acid
Molecular Weight (g/mol): 351.4
Molecular Formula: C21H21NO4
InChi Key: CKLAZLINARHOTG-IBGZPJMESA-N
SMILES: OC(=O)[C@@H]1CCCCN1C(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
Molecular Weight (g/mol) 
  • (7)
  • (3)
Percent Purity 
  • (3)
  • (7)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (7)
  • (3)
  • (7)
  • (7)
  • (3)
  • (3)
  • (7)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)